Myristyltrimethylammonium bromide, 99%
Myristyltrimethylammonium bromide, 99%
Myristyltrimethylammonium bromide, 99%
Myristyltrimethylammonium bromide, 99%
Thermo Scientific Chemicals
Myristyltrimethylammonium bromide, 99%
CAS: 1119-97-7 | C17H38BrN | 336.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA229375000
Price (MYR)
1,163.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,163.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMyristyltrimethylammonium bromide
Name Note99%
CAS1119-97-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
View more
Phase transfer catalyst
RUO – Research Use Only