alpha-Methyl-D-mannopyranoside, 99+%
alpha-Methyl-D-mannopyranoside, 99+%
alpha-Methyl-D-mannopyranoside, 99+%
alpha-Methyl-D-mannopyranoside, 99+%
Thermo Scientific Chemicals
alpha-Methyl-D-mannopyranoside, 99+%
alpha-Methyl-D-mannopyranoside, 99+%, Quantity: 500g, Packaging: Glass bottle, Gray or White, Melting Point: 186.0 deg.C to 194.0 deg.C, Molecular Weight: 194.18, Percent Purity: 99+%, Appearance: (1% aq. soln.) clear, colorless, Assay Percent Range: 99% min. (ELSD) (HPLC) | CAS: 617-04-9 | C7H14O6 | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA229251000
Price (MYR)
589.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
589.00
BOT
Add to cart
Specifications
Chemical Name or Materialα-Methyl-D-mannopyranoside
CAS617-04-9
CAS Min %99.0
CAS Max %100.0
Melting Point186.0°C to 194.0°C