Creatinine, 98%
Creatinine, 98%
Creatinine, 98%
Thermo Scientific Chemicals
Creatinine, 98%
CAS: 60-27-5 | C4H7N3O | 113.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
50 g
Packaging:
Glass bottle
Catalog number FSA228942500
Price (MYR)
1,613.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,613.00
EA
Add to cart
Specifications
Chemical Name or MaterialCreatinine
CAS60-27-5
Melting Point255.0°C
ColorWhite
EINECS Number200-466-7