5-Bromo-2'-deoxyuridine, 99+%
5-Bromo-2'-deoxyuridine, 99+%
5-Bromo-2'-deoxyuridine, 99+%
5-Bromo-2'-deoxyuridine, 99+%
Thermo Scientific Chemicals
5-Bromo-2'-deoxyuridine, 99+%
CAS: 59-14-3 | C9H11BrN2O5 | 307.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
2.5 g
10 g
Packaging:
Glass bottle
Catalog number FSA228595000
Price (MYR)
498.00
BOT
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
498.00
BOT
Add to cart
Specifications
Chemical Name or Material5-Bromo-2′-deoxyuridine
CAS59-14-3
Absorbance0.47 to 0.53 (A250/A260),1.87 to 1.93 (A280/A260),1.45 to 1.55 (A290/A260)
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning