3-Isobutyl-1-methylxanthine, 99+%
3-Isobutyl-1-methylxanthine, 99+%
3-Isobutyl-1-methylxanthine, 99+%
3-Isobutyl-1-methylxanthine, 99+%
Thermo Scientific Chemicals
3-Isobutyl-1-methylxanthine, 99+%
CAS: 28822-58-4 | C10H14N4O2 | 222.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA228420050
Price (MYR)
3,775.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,775.00
EA
Add to cart
Specifications
Chemical Name or Material3-Isobutyl-1-methylxanthine
CAS28822-58-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0