Methyl Violet 2B, pure, indicator grade
Methyl Violet 2B, pure, indicator grade
Methyl Violet 2B, pure, indicator grade
Methyl Violet 2B, pure, indicator grade
Thermo Scientific Chemicals
Methyl Violet 2B, pure, indicator grade
CAS: 8004-87-3 | C24H28ClN3
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA228371000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl violet 2B
Melting Point137.0°C
CAS8004-87-3
Absorption Ratio1 to 1.25
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
View more
Indicator
RUO – Research Use Only