ADA, 99%, for biochemistry
ADA, 99%, for biochemistry
ADA, 99%, for biochemistry
ADA, 99%, for biochemistry
Thermo Scientific Chemicals
ADA, 99%, for biochemistry
CAS: 26239-55-4 | C6H10N2O5 | 190.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA228011000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Absorbance(9% soln. in 1 M NaOH), (9% soln. in 1 M NaOH) at 0nm, 0.08 max. at 300nm
Chemical Name or MaterialADA
Absorbance Conditions(9% soln. in 1M NaOH)
Name NoteFor biochemistry
CAS26239-55-4
View more
Biological buffer
RUO – Research Use Only