ACES, 99%, extra pure
ACES, 99%, extra pure
ACES, 99%, extra pure
ACES, 99%, extra pure
Thermo Scientific Chemicals
ACES, 99%, extra pure
CAS: 7365-82-4 | C4H10N2O4S | 182.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA228001000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Absorbance(c=5, H2O), (c=5, H2O) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
Chemical Name or MaterialACES
Absorbance Conditions(c=5, H2O)
Name NoteExtra Pure
CAS7365-82-4
View more
Biological buffer
RUO – Research Use Only