Chloramphenicol, 98%
Chloramphenicol, 98%
Chloramphenicol, 98%
Chloramphenicol, 98%
Thermo Scientific Chemicals
Chloramphenicol, 98%
Chloramphenicol, CAS # 56-75-7, is a preparation of the broad-spectrum, semisynthetic antibiotic.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Catalog number FSA227920250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialChloramphenicol
TypeChloramphenicol
CAS56-75-7
Health Hazard 2GHS H Statement:
May cause cancer.
Melting Point149.0°C to 153.0°C
View more
General description
  • Chloramphenicol is an organochloride compound that contains a nitrobenzene ring
  • Chloramphenicol acts as a bacteriostatic by binding the 50S ribosomal subunit and preventing peptide formation
  • Stops peptidyltransferase activity by complexing to 50S subunit
Applications
  • Intended for molecular and microbiology applications such as antibiotic resistance gene testing and selective growth experiments and as a selection agent in bacterial cell culture
  • For research use only
RUO – Research Use Only