D(-)-Glutamic acid, 99+%
D(-)-Glutamic acid, 99+%
D(-)-Glutamic acid, 99+%
D(-)-Glutamic acid, 99+%
Thermo Scientific Chemicals
D(-)-Glutamic acid, 99+%
CAS: 6893-26-1 | C5H9NO4 | 147.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA227890250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(-)-Glutamic acid
Name Note99+%
CAS6893-26-1
Melting Point200.0°C to 202.0°C