Pararosaniline chloride, pure, high purity biological stain
CAS: 569-61-9 | C19H18ClN3 | 323.824 g/mol
Have Questions?
Change view
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA227881000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPararosaniline chloride
Melting Point268.0°C to 270.0°C
CAS569-61-9
Health Hazard 1Danger
Health Hazard 2GHS H Statement: May cause cancer.