Cinchonine, 99%
Cinchonine, 99%
Cinchonine, 99%
Thermo Scientific Chemicals
Cinchonine, 99%
CAS: 118-10-5 | C19H22N2O | 294.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA227815000
Price (MYR)
3,796.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,796.00
EA
Add to cart
Specifications
Chemical Name or MaterialCinchonine
CAS118-10-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If s