(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
(+)-5-Fluoro-2'-deoxyuridine, 99+%
Thermo Scientific Chemicals
(+)-5-Fluoro-2'-deoxyuridine, 99+%
CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Packaging:
Glass bottle
Catalog number FSA227605000
Price (MYR)
3,038.00
BOT
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,038.00
BOT
Add to cart
Specifications
Chemical Name or Material(+)-5-Fluoro-2′-deoxyuridine
CAS50-91-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Health Hazard 3GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
IF ON SKIN: Wash with plenty of soap and