Polyvinylpyrrolidone, average M.W. 1.300.000, K85-95
CAS: 9003-39-8 | (C6H9NO)n | 111.14 g/mol
Have Questions?
Change view
Quantity:
500 g
25 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA227475000
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyvinylpyrrolidone
ColorCream-White to White
Name NoteAverage M.W. 1.300.000, K85-95
pH4 to 7 (5% aq. soln.)
CAS9003-39-8