(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
(+)-Griseofulvin, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA227450050
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material(+)-Griseofulvin
Name Note97%
CAS126-07-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger