(-)-Erythromycin, 98%
(-)-Erythromycin, 98%
(-)-Erythromycin, 98%
(-)-Erythromycin, 98%
Thermo Scientific Chemicals
(-)-Erythromycin, 98%
Erythromycin (E-Mycin or erythrocin), CAS # 114-07-8, is a broad-spectrum antibiotic.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Plastic Bottle
Catalog number FSA227330050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or Material(-)-Erythromycin
Type(-)-Erythromycin
Name Note98%
Physical FormPowder
CAS114-07-8
View more
General Description
• Erythromycin is a macrolide antibiotic that works by binding the 50S ribosomal subunit and inhibiting polypeptide formation
• Erythromycin was isolated from Saccharopolyspora erythraea
Applications
• Erythromycin can be used to disrupt the growth of gram-positive and gram-negative bacteria
• Erythromycin has been shown to have motor effects in the gastrointestinal system and can be used for laboratory research on migrating motor complex activity
RUO – Research Use Only
General References:
  1. Dreno, B.; Martin, R.; Moyal, D.; Henley, J.B.; Khammari, A.; Seité, S. Skin microbiome and acne vulgaris: Staphylococcus, a new actor in acne. Exp Dermatol. 2017, 26(9), 798-803.
  2. Wu, F.; Tokach, M.D.; DeRouchey, J.M.; Dritz, S.S.; Woodworth, J.C.; Goodband, R.D.; Chitakasempornkul, K.; Bello, N.M.; Capps, K.; Remfry, S.; Scott, H.M.; Nagaraja, T.G.; Apley, M.D.; Amachawadi, R.G. Effects of Tylosin Administration Routes on the Prevalence of Antimicrobial Resistance Among Fecal Enterococci of Finishing Swine. Foodborne Pathog Dis. 2019, 16(5), 309-316.