Methyl Viologen hydrate, 98%
Methyl Viologen hydrate, 98%
Methyl Viologen hydrate, 98%
Methyl Viologen hydrate, 98%
Methyl Viologen hydrate, 98%
CAS: 1910-42-5 | C12H14Cl2N2 | 257.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA227320050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl Viologen hydrate
Melting Point>300.0°C
Name Note98%
CAS1910-42-5
Health Hazard 1Danger