1-Naphthyl acetate, 99%, Thermo Scientific Chemicals
1-Naphthyl acetate, 99%, Thermo Scientific Chemicals
1-Naphthyl acetate, 99%, Thermo Scientific Chemicals
1-Naphthyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Naphthyl acetate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA227250250
Price (MYR)
905.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
905.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Naphthyl acetate
Name Note99+%
CAS830-81-9
CAS Min %99.0
CAS Max %100.0