2'-Deoxyguanosine hydrate, 99+%
2'-Deoxyguanosine hydrate, 99+%
2'-Deoxyguanosine hydrate, 99+%
2'-Deoxyguanosine hydrate, 99+%
Thermo Scientific Chemicals
2'-Deoxyguanosine hydrate, 99+%
CAS: 961-07-9 | C10H13N5O4 | 267.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
5 g
Catalog number FSA226731000
Price (MYR)
457.00
BOT
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (MYR)
457.00
BOT
Add to cart
Specifications
Chemical Name or Material2′-Deoxyguanosine hydrate
Name Note99+%
CAS961-07-9
Absorbance1.12 to 1.18 (A250/A260) 0.65 to 0.69 (A280/A260)
CAS Min %99.0