2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
2'-Deoxycytidine, 99+%
Thermo Scientific Chemicals
2'-Deoxycytidine, 99+%
CAS: 951-77-9 | C9H13N3O4 | 227.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA226720250
Price (MYR)
6,104.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,104.00
EA
Add to cart
Specifications
Chemical Name or Material2'-Deoxycytidine
CAS951-77-9
CAS Min %99.0
CAS Max %100.0
Melting Point209°C to 211°C