5-Azacytidine, 99%
5-Azacytidine, 99%
5-Azacytidine, 99%
Thermo Scientific Chemicals
5-Azacytidine, 99%
5-Azacytidine, CAS # 320-67-2, is an chemical analog of cytidine that acts as an inhibitor of DNA methyltransferase activity. | CAS: 320-67-2 | C8H12N4O5 | 244.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
1 g
Catalog number FSA226620500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mg
Request bulk or custom format
Specifications
Chemical Name or Material5-Azacytidine
Name Note99%
CAS320-67-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
View more
General Description
  • 5-Azacytidine is a chemical analog of the nucleoside cytidine that incorporates into DNA and inhibits DNA methylation
  • At low doses, 5-azacytidine irreversibly binds the enzyme DNA methyltransferase (DNMT)
Applications
  • 5-Azacytidine has been used for cardiomyocyte differentiation from mesenchymal stem cells
  • It has been used as a potent DNMT inhibitor in the study of tumor cell proliferation and induction of gene expression following demethylation
  • 5-Azacytidine can globally change histone modification patterns
RUO – Research Use Only
General References:
  1. Christman, J.K. 5-Azacytidine and 5-aza-2'-deoxycytidine as inhibitors of DNA methylation: mechanistic studies and their implications for cancer therapy. Oncogene. 2002 Aug 12;21(35):5483-95.
  2. Qian, Q.; Qian, H.; Zhang, X.; Zhu, W.; Yan, Y.; Ye, S.; Peng, X.; Li , W.; Xu, Z.; Sun, L.; Xu ,W. 5-Azacytidine induces cardiac differentiation of human umbilical cord-derived mesenchymal stem cells by activating extracellular regulated kinase. Stem Cells Dev. 2012 Jan;21(1):67-75.
  3. Sato, T.; Issa, J.J.; Kropf,, P. DNA Hypomethylating Drugs in Cancer Therapy. Cold Spring Harb Perspect Med. 2017 May 1, 7(5), a026948.
  4. Komashko, V.M.; Farnham, P.J. 5-azacytidine treatment reorganizes genomic histone modification patterns. Epigenetics. 2010, 5(3), 229-40.
  5. Ebelt, N.D.; Zuniga, E.; Johnson, B.L.; Diamond, D.J.; Manuel, E.R. 5-Azacytidine Potentiates Anti-tumor Immunity in a Model of Pancreatic Ductal Adenocarcinoma. Front Immunol. 2020, 11:538.