Guanosine 5'-monophosphate, disodium salt hydrate, 97%
Guanosine 5'-monophosphate, disodium salt hydrate, 97%
Guanosine 5'-monophosphate, disodium salt hydrate, 97%
Guanosine 5'-monophosphate, disodium salt hydrate, 97%
Guanosine 5'-monophosphate, disodium salt hydrate, 97%
CAS: 5550-12-9 | C10H12N5Na2O8P | 407.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA226601000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialGuanosine 5′-monophosphate,disodium salt hydrate
CAS5550-12-9
Melting Point>300°C
ColorWhite
EINECS Number226-914-1