Kinetin, 99%
Kinetin, 99%
Kinetin, 99%
Thermo Scientific Chemicals
Kinetin, 99%
CAS: 525-79-1 | C10H9N5O | 215.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA226500250
Price (MYR)
5,787.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,787.00
EA
Add to cart
Specifications
Chemical Name or MaterialKinetin
CAS525-79-1
Health Hazard 1Warning
Melting Point269.0°C to 271.0°C
ColorWhite to Beige