6-Benzylaminopurine, 99%
6-Benzylaminopurine, 99%
6-Benzylaminopurine, 99%
6-Benzylaminopurine, 99%
Thermo Scientific Chemicals
6-Benzylaminopurine, 99%
CAS: 1214-39-7 | C12H11N5 | 225.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA226410250
Price (MYR)
1,971.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,971.00
EA
Add to cart
Specifications
Chemical Name or Material6-Benzylaminopurine
CAS1214-39-7
CAS Min %98.5
Health Hazard 1Warning
CAS Max %100.0