Uridine 5'-monophosphate, disodium salt, hydrate, 98%
Uridine 5'-monophosphate, disodium salt, hydrate, 98%
Uridine 5'-monophosphate, disodium salt, hydrate, 98%
Uridine 5'-monophosphate, disodium salt, hydrate, 98%
Thermo Scientific Chemicals
Uridine 5'-monophosphate, disodium salt, hydrate, 98%
homogeneous | CAS: 3387-36-8 | C9H11N2Na2O9P | 368.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA226290050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialUridine 5′-monophosphate, disodium salt, hydrate
CAS3387-36-8
Absorbance0.71 to 0.77 (A250/A260),0.38 to 0.44 (A280/A260)
Melting Point208.0°C to 210.0°C