Bacitracin, zinc salt, potency min. 60 Units/mg
Bacitracin, zinc salt, potency min. 60 Units/mg
Bacitracin, zinc salt, potency min. 60 Units/mg
Bacitracin, zinc salt, potency min. 60 Units/mg
Thermo Scientific Chemicals
Bacitracin, zinc salt, potency min. 60 Units/mg
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass Bottle
Catalog number FSA226110010
Price (MYR)
378.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
378.00
EA
Add to cart
Specifications
Chemical Name or MaterialBacitracin, zinc salt
TypeBacitracin Zinc Salt
Name Notepotency min. 60 Units/mg
Physical FormFine Crystalline Powder
CAS1405-89-6