L(+)-Norvaline, 99%
L(+)-Norvaline, 99%
L(+)-Norvaline, 99%
Thermo Scientific Chemicals
L(+)-Norvaline, 99%
CAS: 6600-40-4 | C5H11NO2 | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass Bottle
Catalog number FSA226020050
Price (MYR)
714.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
714.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(+)-Norvaline
CAS6600-40-4
ColorWhite
Infrared SpectrumAuthentic
Assay Percent Range99%