L(-)-Xylose, 99+%
L(-)-Xylose, 99+%
L(-)-Xylose, 99+%
Thermo Scientific Chemicals
L(-)-Xylose, 99+%
CAS: 609-06-3 | C5H10O5 | 150.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA225991000
Price (MYR)
3,226.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,226.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(-)-Xylose
CAS609-06-3
Melting Point150.0°C to 152.0°C
ColorWhite
EINECS Number210-174-1