Isomaltulose hydrate, 98%
Isomaltulose hydrate, 98%
Isomaltulose hydrate, 98%
Isomaltulose hydrate, 98%
Thermo Scientific Chemicals
Isomaltulose hydrate, 98%
CAS: 343336-76-5 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA225955000
Price (MYR)
1,824.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,824.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsomaltulose hydrate
CAS343336-76-5
Melting Point122.0°C to 124.0°C
Assay Percent Range97.5% min. (ELSD) (HPLC)
PackagingGlass bottle