Maltotriose, 93%
Maltotriose, 93%
Maltotriose, 93%
Thermo Scientific Chemicals
Maltotriose, 93%
CAS: 1109-28-0 | C18H32O16 | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
25 g
Catalog number FSA225945000
Price (MYR)
393.00
BOT
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (MYR)
393.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMaltotriose
CAS1109-28-0
Melting Point132.0°C to 135.0°C
ColorWhite
EINECS Number214-174-2