L(-)-Fucose, 97%
L(-)-Fucose, 97%
L(-)-Fucose, 97%
Thermo Scientific Chemicals
L(-)-Fucose, 97%
CAS: 2438-80-4 | C6H12O5 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA225880250
Price (MYR)
3,320.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,320.00
EA
Add to cart
Specifications
Chemical Name or MaterialL(-)-Fucose
Name Note97%
CAS2438-80-4
Melting Point128.0°C to 141.0°C
ColorBeige to Gray-Purple