2-Aminoisobutyric acid, 98+%
2-Aminoisobutyric acid, 98+%
2-Aminoisobutyric acid, 98+%
2-Aminoisobutyric acid, 98+%
Thermo Scientific Chemicals
2-Aminoisobutyric acid, 98+%
CAS: 62-57-7 | C4H9NO2 | 103.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number FSA225761000
Price (MYR)
873.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
873.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Aminoisobutyric acid
Name Note≥98%
CAS62-57-7
CAS Min %99.0
CAS Max %100.0