D(+)-Melezitose monohydrate, 99+%
D(+)-Melezitose monohydrate, 99+%
D(+)-Melezitose monohydrate, 99+%
D(+)-Melezitose monohydrate, 99+%
Thermo Scientific Chemicals
D(+)-Melezitose monohydrate, 99+%
D(+)-Melezitose monohydrate, CAS # 597-12-6, is the D-form hydrate of melezitose, the trisaccharide.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA225640250
Price (MYR)
899.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
899.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD(+)-Melezitose monohydrate
Name Note99+%
CAS10030-67-8
Melting Point160.0°C
ColorWhite
View more
General Description
• Sap-feeding insects such as ants and aphids produce melezitose to maintain osmotic balance
• Melezitose is produced by some plants as an insect attractant and acts as a metabolite
Applications
• Melezitose can be hydrolyzed to glucose and turanose through alpha-glucosidase activity and can be used for the study of enzyme activity
• Melezitose can be used to distinguish certain strains of bacteria; characterization of phenotype based on the presence or absence of certain sugars assists identification
RUO – Research Use Only
General References:
  1. De Bruyn, T.; van Westen, G.J.; Ijzerman, A.P.; Stieger, B.; de Witte, P.; Augustijns, P.F.; Annaert, P.P. Structure-based identification of OATP1B1/3 inhibitors. Mol Pharmacol. 2013 Jun;83(6):1257-67. doi: 10.1124/mol.112.084152. Epub 2013 Apr 9. PMID: 23571415.
  2. Tan, H.Q.; Li, T.T.; Zhu, C.; Zhang, X.Q.; Wu, M.; Zhu, X.F. Parabacteroides chartae sp. nov., an obligately anaerobic species from wastewater of a paper mill. Int J Syst Evol Microbiol. 2012, 62, 2613-2617.