3,4-Dihydroxyphenylacetic acid, 98%
3,4-Dihydroxyphenylacetic acid, 98%
3,4-Dihydroxyphenylacetic acid, 98%
3,4-Dihydroxyphenylacetic acid, 98%
Thermo Scientific Chemicals
3,4-Dihydroxyphenylacetic acid, 98%
CAS: 102-32-9 | C8H8O4 | 168.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA225600250
Price (MYR)
4,723.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,723.00
EA
Add to cart
Specifications
Chemical Name or Material3, 4-Dihydroxyphenylacetic acid
CAS102-32-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0