N,N-Dimethyloctylamine, 97%
N,N-Dimethyloctylamine, 97%
N,N-Dimethyloctylamine, 97%
N,N-Dimethyloctylamine, 97%
Thermo Scientific Chemicals
N,N-Dimethyloctylamine, 97%
CAS: 7378-99-6 | C10H23N | 157.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA225295000
Price (MYR)
837.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
837.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethyloctylamine
Name Note97%
CAS7378-99-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger