Calcium sulfate dihydrate, 98+%, for analysis
Calcium sulfate dihydrate, 98+%, for analysis
Calcium sulfate dihydrate, 98+%, for analysis
Calcium sulfate dihydrate, 98+%, for analysis
Thermo Scientific Chemicals
Calcium sulfate dihydrate, 98+%, for analysis
CAS: 10101-41-4 | CaH4O6S | 172.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA225275000
Price (MYR)
363.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
363.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCalcium sulfate dihydrate
CAS10101-41-4
ColorWhite
Health Hazard 1
Physical FormPowder