3-Aminosalicylic acid, 97%
3-Aminosalicylic acid, 97%
3-Aminosalicylic acid, 97%
3-Aminosalicylic acid, 97%
Thermo Scientific Chemicals
3-Aminosalicylic acid, 97%
CAS: 570-23-0 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
50 mg
Packaging:
Glass bottle
Catalog number FSA225262500
Price (MYR)
788.00
BOT
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
788.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Aminosalicylic acid
Name Note97%
CAS570-23-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning