Trimethylsilyl isocyanate, 94%
Trimethylsilyl isocyanate, 94%
Trimethylsilyl isocyanate, 94%
Trimethylsilyl isocyanate, 94%
Thermo Scientific Chemicals
Trimethylsilyl isocyanate, 94%
CAS: 1118-02-1 | C4H9NOSi | 115.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA224730250
Price (MYR)
1,542.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,542.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTrimethylsilyl isocyanate
Name Note97%
CAS1118-02-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
To