Titanium(IV) ethoxide, 33-35% TiO2
Titanium(IV) ethoxide, 33-35% TiO2
Titanium(IV) ethoxide, 33-35% TiO2
Titanium(IV) ethoxide, 33-35% TiO2
Thermo Scientific Chemicals
Titanium(IV) ethoxide, 33-35% TiO2
CAS: 3087-36-3 | C8H20O4Ti | 228.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA223762500
Price (MYR)
1,201.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,201.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTitanium(IV) ethoxide
CAS3087-36-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
Wash f
View more
Lewis acid
RUO – Research Use Only