Tin(IV) oxide, 99.9%, (trace metal basis)
Tin(IV) oxide, 99.9%, (trace metal basis)
Tin(IV) oxide, 99.9%, (trace metal basis)
Tin(IV) oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Tin(IV) oxide, 99.9%, (trace metal basis)
CAS: 18282-10-5 | O2Sn | 150.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA223685000
Price (MYR)
1,488.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,488.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTin(IV) oxide
Name Note99.9%
CAS18282-10-5
ColorWhite to Gray
Physical FormPowder
View more
For ultra trace analysis
RUO – Research Use Only