Zinc sulfide, 99.99%, (trace metal basis), < 10 microns
Zinc sulfide, 99.99%, (trace metal basis), &lt; 10 microns
Zinc sulfide, 99.99%, (trace metal basis), &lt; 10 microns
Zinc sulfide, 99.99%, (trace metal basis), &lt; 10 microns
Thermo Scientific Chemicals
Zinc sulfide, 99.99%, (trace metal basis), < 10 microns
CAS: 1314-98-3 | SZn | 97.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA223650250
Price (MYR)
394.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
394.00
EA
Add to cart
Specifications
Chemical Name or MaterialZinc sulfide
Name Note99.99%, <10 microns
CAS1314-98-3
ColorWhite to Yellow
Health Hazard 1MOISTURE SENSITIVE
View more
For ultra trace analysis
RUO – Research Use Only