hydrogen tetrachloroaurate(III) hydrate
hydrogen tetrachloroaurate(III) hydrate
hydrogen tetrachloroaurate(III) hydrate
hydrogen tetrachloroaurate(III) hydrate
Thermo Scientific Chemicals
hydrogen tetrachloroaurate(III) hydrate
CAS: 27988-77-8 | AuCl4H | 339.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA223620050
Price (MYR)
6,488.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,488.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydrogen tetrachloroaurate(III) hydrate
Melting Point30.0°C
Name Notep.a.
CAS27988-77-8
ColorOrange