1-(Trimethylsilyl)-1-propyne, 98%, Thermo Scientific Chemicals
1-(Trimethylsilyl)-1-propyne, 98%, Thermo Scientific Chemicals
1-(Trimethylsilyl)-1-propyne, 98%, Thermo Scientific Chemicals
1-(Trimethylsilyl)-1-propyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(Trimethylsilyl)-1-propyne, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA223530050
Price (MYR)
597.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
597.00
BOT
Add to cart
Specifications
Chemical Name or Material1-(Trimethylsilyl)-1-propyne
Name Note98%
CAS6224-91-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger