1-Decyne, 98%
1-Decyne, 98%
1-Decyne, 98%
Thermo Scientific Chemicals
1-Decyne, 98%
CAS: 764-93-2 | C10H18 | 138.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA223430250
Price (MYR)
791.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
791.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Decyne
Name Note98%
CAS764-93-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning