Hydrobromic acid, for analysis, ca. 48% solution in water
Hydrobromic acid, for analysis, ca. 48% solution in water
Hydrobromic acid, for analysis, ca. 48% solution in water
Hydrobromic acid, for analysis, ca. 48% solution in water
Thermo Scientific Chemicals
Hydrobromic acid, for analysis, ca. 48% solution in water
CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA223325000
Price (MYR)
507.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
507.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydrobromic acid
ColorYellow
CAS10035-10-6
Melting Point-11.0°C
Boiling Point126.0°C to 128.0°C
View more
C-X bond formation reagent
RUO – Research Use Only