Sodium chlorite, 80%, pure, unstabilized
Sodium chlorite, 80%, pure, unstabilized
Sodium chlorite, 80%, pure, unstabilized
Sodium chlorite, 80%, pure, unstabilized
Thermo Scientific Chemicals
Sodium chlorite, 80%, pure, unstabilized
Sodium chlorite, 80%, ClNaO2, CAS Number-7758-19-2, 7647-14-5, unii-g538ebv4vf, alcide ld, neo silox d, chlorous acid, sodium salt, caswell no. 755, sodiumchlorite, textone, sodium chlorite, textile, chlorite sodium, 500g, CHEBI:78667, White, 231-836-6, 05,603; 09,423; 11,481; 13,280, 90.44, Pure | CAS: 7758-19-2 | ClNaO2 | 90.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2.5 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number FSA223235000
Price (MYR)
507.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
507.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium chlorite
Melting Point180.0°C to 200.0°C
Name NoteUnstabilized
CAS7758-19-2