Sodium chlorate, 99+%, extra pure
Sodium chlorate, 99+%, extra pure
Sodium chlorate, 99+%, extra pure
Sodium chlorate, 99+%, extra pure
Thermo Scientific Chemicals
Sodium chlorate, 99+%, extra pure
CAS: 7775-09-9 | ClNaO3 | 106.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA223222500
Price (MYR)
224.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
224.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium chlorate
Melting Point248.0°C to 261.0°C
CAS7775-09-9
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only