Sodium perborate tetrahydrate, 97%, pure
Sodium perborate tetrahydrate, 97%, pure
Sodium perborate tetrahydrate, 97%, pure
Sodium perborate tetrahydrate, 97%, pure
Sodium perborate tetrahydrate, 97%, pure
Thermo Scientific Chemicals
Sodium perborate tetrahydrate, 97%, pure
CAS: 10486-00-7 | BNaO3·4H2O | 153.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA223202500
Price (MYR)
329.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
329.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium perborate tetrahydrate
Melting Point60.0°C
CAS10486-00-7
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Suitable for battery materials development.
RUO – Research Use Only