N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA223170250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-(2-Aminoethyl)acetamide
CAS1001-53-2
CAS Min %88.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0