Potassium ferricyanide, 99+%, for analysis
Potassium ferricyanide, 99+%, for analysis
Potassium ferricyanide, 99+%, for analysis
Potassium ferricyanide, 99+%, for analysis
Thermo Scientific Chemicals
Potassium ferricyanide, 99+%, for analysis
CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA223111000
Price (MYR)
308.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
308.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium ferricyanide
Name Notep.a.
CAS13746-66-2
ColorOrange to Red
Health Hazard 1Warning
View more
Indicator
RUO – Research Use Only